missing translation for 'onlineSavingsMsg'
Learn More

trans,trans-Muconic Acid 98.0+%, TCI America™

Catalog No. M04731G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M04731G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M04731G Supplier TCI America Supplier No. M04731G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3588-17-8
Molecular Formula C6H6O4
Molecular Weight (g/mol) 142.11
MDL Number MFCD00002702
InChI Key TXXHDPDFNKHHGW-ZPUQHVIOSA-N
Synonym trans,trans-muconic acid, muconic acid, e,e-muconic acid, 2,4-hexadienedioic acid, trans, trans-muconic acid, hexa-2,4-dienedioic acid, 2e,4e-hexa-2,4-dienedioic acid, 1,3-butadiene-1,4-dicarboxylic acid, 2,4-hexadienedioic acid, e,e, trans,trans-2,4-hexadienedioic acid
PubChem CID 5356793
ChEBI CHEBI:27036
IUPAC Name (2E,4E)-hexa-2,4-dienedioic acid
SMILES C(=CC(=O)O)C=CC(=O)O

Specifications

Quantity 1 g
Formula Weight 142.11
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material trans,trans-Muconic Acid
RTECSNumber MM2235070
TSCA No