missing translation for 'onlineSavingsMsg'
Learn More

trans,trans-4'-Propyl-4-(p-tolyl)bicyclohexyl 98.0+%, TCI America™

Catalog No. p-7135562
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P24041G 1 g
P24045G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P24041G Supplier TCI America Supplier No. P24041G

Chemical Identifiers

CAS 84656-75-7
Molecular Formula C22H34
Molecular Weight (g/mol) 298.51
MDL Number MFCD13182311
InChI Key SDESCXGEQILYTQ-UHFFFAOYSA-N
Synonym 1-methyl-4-trans,trans-4'-propyl 1,1'-bicyclohexyl-4-yl benzene, trans,trans-4'-p-tolyl-4-propyl-bicyclohexyl, benzene, 1-methyl-4-4'-propyl 1,1'-bicyclohexyl-4-yl, trans,trans-4-4-methylphenyl-4'-n-propylbicyclohexyl, 4-trans-4 trans-4-propylcyclohexyl cyclohexyl toluene, trans,trans-4-p-tolyl-4-propyl-bicyclohexyl, trans,trans-4'-propyl-4-p-tolyl bicyclohexyl, 4-propyl-4'-p-tolyl-1,1'-bi cyclohexane, 1-methyl-4-4-4-propylcyclohexyl cyclohexyl benzene, trans,trans-4'-propyl-4-4-methylphenyl bicyclohexyl
PubChem CID 576378
IUPAC Name 4-(4-methylphenyl)-4'-propyl-1,1'-bi(cyclohexane)
SMILES CCCC1CCC(CC1)C1CCC(CC1)C1=CC=C(C)C=C1

Specifications

Melting Point 110°C
Color White
Quantity 1 g
Formula Weight 298.51
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material trans,trans-4′-Propyl-4-(p-tolyl)bicyclohexyl
EINECSNumber (4)-1985
TSCA No