missing translation for 'onlineSavingsMsg'
Learn More

trans,trans-4-Pentyl-4'-vinylbicyclohexyl 98.0+%, TCI America™

Catalog No. P23181G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P23181G 1 g
P23185G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P23181G Supplier TCI America Supplier No. P23181G
Only null left

Chemical Identifiers

CAS 129738-34-7
Molecular Formula C19H34
Molecular Weight (g/mol) 262.48
MDL Number MFCD11053467
InChI Key ALEUVXQGVOQDBD-UHFFFAOYSA-N
PubChem CID 19937987
IUPAC Name 4-ethenyl-4'-pentyl-1,1'-bi(cyclohexane)
SMILES CCCCCC1CCC(CC1)C1CCC(CC1)C=C

Specifications

Melting Point 49°C
Color White
Quantity 1 g
Synonym trans,trans-4-Amyl-4′C-vinylbicyclohexyl
Formula Weight 262.48
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material trans,trans-4-Pentyl-4′-vinylbicyclohexyl
EINECSNumber (4)-1900
TSCA No