missing translation for 'onlineSavingsMsg'
Learn More

trans,trans-4-Butyl-4'-vinylbicyclohexyl 98.0+%, TCI America™

Catalog No. p-7123971
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B51231G 1 g
B51235G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B51231G Supplier TCI America Supplier No. B51231G
Only null left

Chemical Identifiers

CAS 153429-47-1
Molecular Formula C18H32
Molecular Weight (g/mol) 248.454
MDL Number MFCD25541699
InChI Key HNJLLLQZSIESBM-UHFFFAOYSA-N
PubChem CID 19702460
IUPAC Name 1-butyl-4-(4-ethenylcyclohexyl)cyclohexane
SMILES CCCCC1CCC(CC1)C2CCC(CC2)C=C

Specifications

Melting Point 55°C
Color White
Quantity 1 g
Formula Weight 248.45
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material trans,trans-4-Butyl-4′-vinylbicyclohexyl