missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans,trans-2,4-Undecadienal, 90+%, remainder mainly trans,cis isomer
CAS: 30361-29-6 | C11H18O | 166.264 g/mol
$45.90 - $154.30
Chemical Identifiers
| CAS | 30361-29-6 |
|---|---|
| Molecular Formula | C11H18O |
| Molecular Weight (g/mol) | 166.264 |
| MDL Number | MFCD00014677 |
| InChI Key | UVIUIIFPIWRILL-XBLVEGMJSA-N |
| Synonym | 2,4-undecadienal, 2e,4e-undeca-2,4-dienal, undeca-2,4-dien-1-al, trans,trans-2,4-undecadien-1-al, 2,4-undecadienal, 2e,4e, 2,4-trans,trans-undecadienal, unii-7l26s7bw06, 2,4-undecadienal, e,e, trans,trans-2,4-undecadienal, e,e-2,4-undecadien-1-al |
| PubChem CID | 5367531 |
| IUPAC Name | (2E,4E)-undeca-2,4-dienal |
| SMILES | CCCCCCC=CC=CC=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1341906
|
Thermo Scientific Chemicals
A1341906 |
5 g |
Each for $45.90
|
|
|||||
|
AAA1341914
|
Thermo Scientific Chemicals
A1341914 |
25 g |
Each for $154.30
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 30361-29-6 | |
| 166.264 | |
| UVIUIIFPIWRILL-XBLVEGMJSA-N | |
| 5367531 | |
| CCCCCCC=CC=CC=O |
| C11H18O | |
| MFCD00014677 | |
| 2,4-undecadienal, 2e,4e-undeca-2,4-dienal, undeca-2,4-dien-1-al, trans,trans-2,4-undecadien-1-al, 2,4-undecadienal, 2e,4e, 2,4-trans,trans-undecadienal, unii-7l26s7bw06, 2,4-undecadienal, e,e, trans,trans-2,4-undecadienal, e,e-2,4-undecadien-1-al | |
| (2E,4E)-undeca-2,4-dienal |
Specifications
| 30361-29-6 | |
| 128°C to 129°C (10 mmHg) | |
| C11H18O | |
| MFCD00014677 | |
| 1706330 | |
| 2,4-undecadienal, 2e,4e-undeca-2,4-dienal, undeca-2,4-dien-1-al, trans,trans-2,4-undecadien-1-al, 2,4-undecadienal, 2e,4e, 2,4-trans,trans-undecadienal, unii-7l26s7bw06, 2,4-undecadienal, e,e, trans,trans-2,4-undecadienal, e,e-2,4-undecadien-1-al | |
| CCCCCCC=CC=CC=O | |
| 166.264 | |
| 166.27 | |
| trans,trans-2,4-Undecadienal |
| 0.87 | |
| 107°C (225°F) | |
| 1.512 | |
| 5 g | |
| Air sensitive | |
| UVIUIIFPIWRILL-XBLVEGMJSA-N | |
| (2E,4E)-undeca-2,4-dienal | |
| 5367531 | |
| ≥90% | |
| remainder mainly trans,cis isomer |
Safety and Handling
EINECSNumber : 250-148-7
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only