missing translation for 'onlineSavingsMsg'
Learn More

trans,trans-2,4-Undecadienal, 90+%, remainder mainly trans,cis isomer

Catalog No. AAA1341914
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1341914 25 g
AAA1341906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1341914 Supplier Thermo Scientific Chemicals Supplier No. A1341914
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 30361-29-6 | C11H18O | 166.264 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 30361-29-6
Molecular Formula C11H18O
Molecular Weight (g/mol) 166.264
MDL Number MFCD00014677
InChI Key UVIUIIFPIWRILL-XBLVEGMJSA-N
Synonym 2,4-undecadienal, 2e,4e-undeca-2,4-dienal, undeca-2,4-dien-1-al, trans,trans-2,4-undecadien-1-al, 2,4-undecadienal, 2e,4e, 2,4-trans,trans-undecadienal, unii-7l26s7bw06, 2,4-undecadienal, e,e, trans,trans-2,4-undecadienal, e,e-2,4-undecadien-1-al
PubChem CID 5367531
IUPAC Name (2E,4E)-undeca-2,4-dienal
SMILES CCCCCCC=CC=CC=O

Specifications

Density 0.87
Boiling Point 128°C to 129°C (10 mmHg)
Flash Point 107°C (225°F)
Refractive Index 1.512
Quantity 25 g
Beilstein 1706330
Sensitivity Air sensitive
Formula Weight 166.27
Percent Purity ≥90%
Assay remainder mainly trans,cis isomer
Chemical Name or Material trans,trans-2,4-Undecadienal
Show More Show Less
EINECSNumber 250-148-7
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only