missing translation for 'onlineSavingsMsg'
Learn More

trans,trans-2,4-Hexadien-1-ol, 99%, stabilized

Catalog No. AC325572500
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC325572500 250 g
AC325570100 10 g
AC325570500 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC325572500 Supplier Thermo Scientific Chemicals Supplier No. 325572500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 17102-64-6 | C6H10O | 98.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 17102-64-6
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.15
MDL Number MFCD00002925
InChI Key MEIRRNXMZYDVDW-MQQKCMAXSA-N
Synonym sorbic alcohol, 2,4-hexadien-1-ol, sorbyl alcohol, trans,trans-2,4-hexadien-1-ol, 2,4-hexadienol, hexacose, hexakose, sorbinic alcohol, 1-hydroxy-2,4-hexadiene, 2e,4e-hexa-2,4-dien-1-ol
PubChem CID 641256
IUPAC Name (2E,4E)-hexa-2,4-dien-1-ol
SMILES C\C=C\C=C\CO

Specifications

Melting Point 30.5°C to 31.5°C
Color Colorless to Yellow
Density 0.8700g/mL
Boiling Point 80.0°C (12.0 mmHg)
Flash Point 72°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Refractive Index 1.4990 to 1.5010
Linear Formula CH3CH=CHCH=CHCH2OH
Quantity 250 g
Merck Index 15,8848
Specific Gravity 0.87
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,ether and oils
Stabilizer 0.1% alpha-tocopherol
Formula Weight 98.14
Percent Purity 99%
Physical Form Liquid After Melting
Chemical Name or Material trans, trans-2, 4-Hexadien-1-ol
Show More Show Less
Hazard Category GHS Signal Word: Danger
EINECSNumber 241-173-4

RUO – Research Use Only