missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans,trans-2,4-Dodecadienal, 90+%, remainder mainly trans, cis isomer
CAS: 21662-16-8 | C12H20O | 180.29 g/mol
$82.62 - $229.49
Chemical Identifiers
| CAS | 21662-16-8 |
|---|---|
| Molecular Formula | C12H20O |
| Molecular Weight (g/mol) | 180.29 |
| MDL Number | MFCD00014678 |
| InChI Key | QKTZBZWNADPFOL-BNFZFUHLSA-N |
| Synonym | 2e,4e-dodeca-2,4-dienal, 2,4-dodecadienal, e,e-2,4-dodecadienal, trans,trans-2,4-dodecadienal, 2,4-dodecadien-1-al, 2e,4e-2,4-dodecadienal, e,e-2,4-dodecadien-1-al, 2,4-dodecadienal, e,e, unii-vo340rj9lu |
| PubChem CID | 5367530 |
| IUPAC Name | (2E,4E)-dodeca-2,4-dienal |
| SMILES | CCCCCCC\C=C\C=C\C=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL0600203
|
Thermo Scientific Chemicals
L0600203 |
1 g |
N/A
|
|
|||||
|
AAL0600206
|
Thermo Scientific Chemicals
L0600206 |
5 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 21662-16-8 | |
| 180.29 | |
| QKTZBZWNADPFOL-BNFZFUHLSA-N | |
| 5367530 | |
| CCCCCCC\C=C\C=C\C=O |
| C12H20O | |
| MFCD00014678 | |
| 2e,4e-dodeca-2,4-dienal, 2,4-dodecadienal, e,e-2,4-dodecadienal, trans,trans-2,4-dodecadienal, 2,4-dodecadien-1-al, 2e,4e-2,4-dodecadienal, e,e-2,4-dodecadien-1-al, 2,4-dodecadienal, e,e, unii-vo340rj9lu | |
| (2E,4E)-dodeca-2,4-dienal |
Specifications
| 21662-16-8 | |
| 130°C (6 mmHg) | |
| C12H20O | |
| MFCD00014678 | |
| 1708096 | |
| QKTZBZWNADPFOL-BNFZFUHLSA-N | |
| (2E,4E)-dodeca-2,4-dienal | |
| 5367530 | |
| ≥90% | |
| remainder mainly trans, cis isomer |
| 0.863 | |
| 104°C (219°F) | |
| 1.51 | |
| 1 g | |
| 2e,4e-dodeca-2,4-dienal, 2,4-dodecadienal, e,e-2,4-dodecadienal, trans,trans-2,4-dodecadienal, 2,4-dodecadien-1-al, 2e,4e-2,4-dodecadienal, e,e-2,4-dodecadien-1-al, 2,4-dodecadienal, e,e, unii-vo340rj9lu | |
| CCCCCCC\C=C\C=C\C=O | |
| 180.29 | |
| 180.29 | |
| trans,trans-2,4-Dodecadienal |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
EINECSNumber : 244-517-1
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only