missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans,trans-2,4-Decadien-1-ol, 90%, remainder mainly trans, cis isomer
CAS: 18409-21-7 | C10H18O | 154.25 g/mol
$990.34 - $990.34
Chemical Identifiers
| CAS | 18409-21-7 |
|---|---|
| Molecular Formula | C10H18O |
| Molecular Weight (g/mol) | 154.25 |
| MDL Number | MFCD00014052 |
| InChI Key | NUBWFSDCZULDCI-BLHCBFLLSA-N |
| Synonym | 2,4-decadien-1-ol, 2e,4e-2,4-decadien-1-ol, 2e,4e-deca-2,4-dien-1-ol, e,e-2,4-decadien-1-ol, unii-21kkw523r4, trans,trans-2,4-decadien-1-ol, trans,trans-2,4-decadienol, 2,4-decadien-1-ol, 2e,4e, trans-2,4-decadienol, 2,4-decadien-1-ol, e,e |
| PubChem CID | 5362696 |
| IUPAC Name | (2E,4E)-deca-2,4-dien-1-ol |
| SMILES | CCCCC\C=C\C=C\CO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1598914
|
Thermo Scientific Chemicals
A1598914 |
25 g |
Each for $990.34
|
|
|||||
Description
trans,trans-2,4-Decadien-1-ol is used as food flavor and as an intermediate in organic chemical synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationstrans,trans-2,4-Decadien-1-ol is used as food flavor and as an intermediate in organic chemical synthesis.
Solubility
Not miscible in water. Soluble in fat.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
| 18409-21-7 | |
| 154.25 | |
| NUBWFSDCZULDCI-BLHCBFLLSA-N | |
| 5362696 | |
| CCCCC\C=C\C=C\CO |
| C10H18O | |
| MFCD00014052 | |
| 2,4-decadien-1-ol, 2e,4e-2,4-decadien-1-ol, 2e,4e-deca-2,4-dien-1-ol, e,e-2,4-decadien-1-ol, unii-21kkw523r4, trans,trans-2,4-decadien-1-ol, trans,trans-2,4-decadienol, 2,4-decadien-1-ol, 2e,4e, trans-2,4-decadienol, 2,4-decadien-1-ol, e,e | |
| (2E,4E)-deca-2,4-dien-1-ol |
Specifications
| 18409-21-7 | |
| 80°C (0.5 mmHg) | |
| C10H18O | |
| MFCD00014052 | |
| 2,4-decadien-1-ol, 2e,4e-2,4-decadien-1-ol, 2e,4e-deca-2,4-dien-1-ol, e,e-2,4-decadien-1-ol, unii-21kkw523r4, trans,trans-2,4-decadien-1-ol, trans,trans-2,4-decadienol, 2,4-decadien-1-ol, 2e,4e, trans-2,4-decadienol, 2,4-decadien-1-ol, e,e | |
| NUBWFSDCZULDCI-BLHCBFLLSA-N | |
| (2E,4E)-deca-2,4-dien-1-ol | |
| 5362696 | |
| 90% | |
| remainder mainly trans, cis isomer |
| 0.87 | |
| >110°C (230°F) | |
| 1.489 | |
| 25 g | |
| Not miscible in water. Soluble in fat. | |
| CCCCC\C=C\C=C\CO | |
| 154.25 | |
| 154.26 | |
| trans,trans-2,4-Decadien-1-ol |
Safety and Handling
EINECSNumber : 242-291-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only