missing translation for 'onlineSavingsMsg'
Learn More

trans,trans-1,3-Butadiene-1,4-dicarboxylic acid, 98%

Catalog No. aal0398706
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0398706 5 g
AAL0398703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0398706 Supplier Thermo Scientific Chemicals Supplier No. L0398706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3588-17-8 | C6H6O4 | 142.11 g/mol

trans,trans-1,3-Butadiene-1,4-dicarboxylic acid is a metabolite found in urine that determines biological exposure index for workers exposed to benzene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans,trans-1,3-Butadiene-1,4-dicarboxylic acid is a metabolite found in urine that determines biological exposure index for workers exposed to benzene.

Solubility
Soluble in dimethyl sulfoxide and methanol.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 3588-17-8
Molecular Formula C6H6O4
Molecular Weight (g/mol) 142.11
MDL Number MFCD00002702
InChI Key TXXHDPDFNKHHGW-ZPUQHVIOSA-N
Synonym trans,trans-muconic acid, muconic acid, e,e-muconic acid, 2,4-hexadienedioic acid, trans, trans-muconic acid, hexa-2,4-dienedioic acid, 2e,4e-hexa-2,4-dienedioic acid, 1,3-butadiene-1,4-dicarboxylic acid, 2,4-hexadienedioic acid, e,e, trans,trans-2,4-hexadienedioic acid
PubChem CID 5356793
ChEBI CHEBI:27036
IUPAC Name (2E,4E)-hexa-2,4-dienedioic acid
SMILES C(=CC(=O)O)C=CC(=O)O

Specifications

Melting Point ∽300°C (decomposition)
Quantity 5 g
Beilstein 1722248
Merck Index 14,6299
Solubility Information Soluble in dimethyl sulfoxide and methanol.
Formula Weight 142.11
Percent Purity ≥98%
Chemical Name or Material trans,trans-1,3-Butadiene-1,4-dicarboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 222-724-8
RTECSNumber MM2235070
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only