missing translation for 'onlineSavingsMsg'
Learn More

trans-Stilbene Oxide 98.0+%, TCI America™

Catalog No. p-7135851
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S02625G 5 g
S026225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S02625G Supplier TCI America Supplier No. S02625G
Only null left

Chemical Identifiers

CAS 1439-07-2
Molecular Formula C14H12O
Molecular Weight (g/mol) 196.249
MDL Number MFCD00064311
InChI Key ARCJQKUWGAZPFX-KWCCSABGSA-N
Synonym trans-stilbene oxide, 3r-2,3-diphenyloxirane, 2r-2alpha,3-diphenyloxirane
PubChem CID 5742860
IUPAC Name (3R)-2,3-diphenyloxirane
SMILES C1=CC=C(C=C1)C2C(O2)C3=CC=CC=C3

Specifications

Melting Point 68°C
Color White-Yellow
Quantity 5 g
Formula Weight 196.25
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material trans-Stilbene Oxide