missing translation for 'onlineSavingsMsg'
Learn More

trans-Stilbene, 98%

Catalog No. p-7080628
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1023222 100 g
AAA1023236 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1023222 Supplier Thermo Scientific Chemicals Supplier No. A1023222

CAS: 103-30-0 | C14H12 | 180.25 g/mol

trans-Stilbene is used in the manufacturing of dye lasers, optical brighteners, non-steroidal synthetic estrogens such as diethylstilbestrol, fosfestrol and dienestrol. It is used as a phosphor and a scintillator. It is also involved in transition metal catalyzed asymmetric epoxidation and dihydroxylation. Further, it is used as raw material in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-Stilbene is used in the manufacturing of dye lasers, optical brighteners, non-steroidal synthetic estrogens such as diethylstilbestrol, fosfestrol and dienestrol. It is used as a phosphor and a scintillator. It is also involved in transition metal catalyzed asymmetric epoxidation and dihydroxylation. Further, it is used as raw material in organic synthesis.

Solubility
Soluble in benzene and ether. Insoluble in water.

Notes
Air and light sensitive. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 103-30-0
Molecular Formula C14H12
Molecular Weight (g/mol) 180.25
MDL Number MFCD00064300
InChI Key PJANXHGTPQOBST-VAWYXSNFSA-N
Synonym trans-stilbene, e-stilbene, stilbene, 1,2-diphenylethylene, trans-1,2-diphenylethylene, bibenzylidene, bibenzylidine, bibenzal, trans-1,2-diphenylethene, alpha,beta-diphenylethylene
PubChem CID 638088
ChEBI CHEBI:36007
IUPAC Name (E)-stilbene
SMILES C1=CC=C(C=C1)C=CC2=CC=CC=C2

Specifications

Melting Point 122°C to 125°C
Density 1.14
Boiling Point 305°C to 307°C
Odor Odorless
Quantity 100 g
UN Number UN3077
Beilstein 1616740
Merck Index 14,8817
Solubility Information Soluble in benzene and ether. Insoluble in water.
Formula Weight 180.25
Percent Purity 98%
Chemical Name or Material trans-Stilbene
Show More Show Less
Hazard Category H302-H319
Hazard Statement GHS H Statement
H302-H319
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 203-098-5
RTECSNumber WJ4926500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only