missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-p-Coumaric Acid 98.0+%, TCI America™
Supplier: TCI America C0393100G
Specifications
| trans-p-Coumaric Acid | |
| 213°C | |
| C9H8O3 | |
| 100 g | |
| NGSWKAQJJWESNS-ZZXKWVIFSA-N | |
| (E)-3-(4-hydroxyphenyl)prop-2-enoic acid | |
| 637542 | |
| 164.16 | |
| Crystalline Powder |
| 501-98-4 | |
| White-Yellow | |
| MFCD00004399 | |
| p-coumaric acid, 4-hydroxycinnamic acid, p-hydroxycinnamic acid, 4-coumaric acid, trans-4-hydroxycinnamic acid, trans-p-coumaric acid, p-cumaric acid, 3-4-hydroxyphenyl acrylic acid, para-coumaric acid, hydroxycinnamic acid | |
| C1=CC(=CC=C1C=CC(=O)O)O | |
| 164.16 | |
| CHEBI:32374 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 501-98-4 | |
| 164.16 | |
| NGSWKAQJJWESNS-ZZXKWVIFSA-N | |
| 637542 | |
| (E)-3-(4-hydroxyphenyl)prop-2-enoic acid |
| C9H8O3 | |
| MFCD00004399 | |
| p-coumaric acid, 4-hydroxycinnamic acid, p-hydroxycinnamic acid, 4-coumaric acid, trans-4-hydroxycinnamic acid, trans-p-coumaric acid, p-cumaric acid, 3-4-hydroxyphenyl acrylic acid, para-coumaric acid, hydroxycinnamic acid | |
| CHEBI:32374 | |
| C1=CC(=CC=C1C=CC(=O)O)O |
Safety and Handling
RTECSNumber : GD9095000
TSCA : No