missing translation for 'onlineSavingsMsg'
Learn More

trans-p-Coumaric Acid 98.0+%, TCI America™

Catalog No. C039325G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C039325G 25 g
C0393100G 100 g
C0393500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C039325G Supplier TCI America Supplier No. C039325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 501-98-4
Molecular Formula C9H8O3
Molecular Weight (g/mol) 164.16
MDL Number MFCD00004399
InChI Key NGSWKAQJJWESNS-ZZXKWVIFSA-N
Synonym p-coumaric acid, 4-hydroxycinnamic acid, p-hydroxycinnamic acid, 4-coumaric acid, trans-4-hydroxycinnamic acid, trans-p-coumaric acid, p-cumaric acid, 3-4-hydroxyphenyl acrylic acid, para-coumaric acid, hydroxycinnamic acid
PubChem CID 637542
ChEBI CHEBI:32374
IUPAC Name (E)-3-(4-hydroxyphenyl)prop-2-enoic acid
SMILES C1=CC(=CC=C1C=CC(=O)O)O

Specifications

Melting Point 213°C
Color White-Yellow
Quantity 25 g
Formula Weight 164.16
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material trans-p-Coumaric Acid
RTECSNumber GD9095000
TSCA No