missing translation for 'onlineSavingsMsg'
Learn More

trans-N,N'-Dimethylcyclohexane-1,2-diamine 98.0+%, TCI America™

Catalog No. p-7127577
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D29461G 1 g
D29465G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D29461G Supplier TCI America Supplier No. D29461G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 67579-81-1
Molecular Formula C8H18N2
Molecular Weight (g/mol) 142.246
MDL Number MFCD03001702
InChI Key JRHPOFJADXHYBR-HTQZYQBOSA-N
Synonym 1r,2r-n,n'-dimethyl-1,2-cyclohexanediamine, 1r,2r-n1,n2-dimethylcyclohexane-1,2-diamine, 1r,2r---n,n'-dimethylcyclohexane-1,2-diamine, trans-n1,n2-dimethylcyclohexane-1,2-diamine, trans-1r,2r-n,n'-bismethyl-1,2-cyclohexanediamine, trans-n,n'-dimethylcyclohexane-1,2-diamine, trans-n,n'-dimethyl-1,2-cyclohexanediamine, trans-1,2-bis methylamino cyclohexane, trans-1r,2r-n,n'-bismethyl-1,2-cyclohexane diamine, 1r,2r---1,2-bis methylamino cyclohexane
PubChem CID 2733821
IUPAC Name (1R,2R)-1-N,2-N-dimethylcyclohexane-1,2-diamine
SMILES CNC1CCCCC1NC

Specifications

Color Yellow
Boiling Point 83°C
Quantity 1 g
UN Number 2735
Formula Weight 142.25
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material trans-N,N′-Dimethylcyclohexane-1,2-diamine