missing translation for 'onlineSavingsMsg'
Learn More

trans-N,N'-Diacetylcyclohexane-1,2-diamine 98.0+%, TCI America™

Catalog No. D32265G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D32265G 5 g
D322625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D32265G Supplier TCI America Supplier No. D32265G
Only null left

Chemical Identifiers

CAS 70924-78-6
Molecular Formula C10H18N2O2
Molecular Weight (g/mol) 198.266
InChI Key MZYLAHHTXWRMOL-NXEZZACHSA-N
Synonym trans-1,2-Bis(acetamido)cyclohexane
PubChem CID 11805689
IUPAC Name N-[(1R,2R)-2-acetamidocyclohexyl]acetamide
SMILES CC(=O)NC1CCCCC1NC(=O)C

Specifications

Melting Point 270°C
Color White-Yellow
Quantity 5 g
Formula Weight 198.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material trans-N,N′-Diacetylcyclohexane-1,2-diamine