missing translation for 'onlineSavingsMsg'
Learn More

trans-N-Fmoc-4-tert-butoxy-L-proline, 98%

Catalog No. AAH6379103
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6379103 1 g
AAH6379106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6379103 Supplier Thermo Scientific Chemicals Supplier No. H6379103
Only null left

CAS: 122996-47-8 | C24H27NO5 | 409.482 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 122996-47-8
Molecular Formula C24H27NO5
Molecular Weight (g/mol) 409.482
MDL Number MFCD00151930
InChI Key WPBXBYOKQUEIDW-FXMQYSIJSA-N
Synonym fmoc-4-tert-butoxy-l-proline, 2s-4-tert-butoxy-1-9h-fluoren-9-ylmethoxy carbonyl pyrrolidine-2-carboxylic acid, fmoc-o-tert-butyl-l-hydroxyproline, ksc491o9t, n-alpha-9-fluorenylmethyloxycarbonyl-o-t-butyl-trans-l-hydroxyproline, 2s-1-9h-fluoren-9-ylmethoxycarbonyl-4-2-methylpropan-2-yl oxy pyrrolidine-2-carboxylic acid
PubChem CID 6916162
IUPAC Name (2S)-1-(9H-fluoren-9-ylmethoxycarbonyl)-4-[(2-methylpropan-2-yl)oxy]pyrrolidine-2-carboxylic acid
SMILES CC(C)(C)OC1CC(N(C1)C(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24)C(=O)O

Specifications

Color White
Beilstein 6161467
Quantity 1 g
Formula Weight 409.47
Percent Purity 98%
Physical Form Powder
Chemical Name or Material trans-N-Fmoc-4-tert-butoxy-L-proline
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only