missing translation for 'onlineSavingsMsg'
Learn More

trans-N-(6,6-Dimethyl-2-hepten-4-ynyl)-N-methyl-1-naphthylmethylamine Hydrochloride 98.0+%, TCI America™

Catalog No. p-7127085
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
D20491G 1 g
D20495G 5 g
2 options

Catalog No. D20491G

Supplier: TCI America D20491G

Only null left

Chemical Identifiers

CAS 78628-80-5
Molecular Formula C21H26ClN
Molecular Weight (g/mol) 327.896
MDL Number MFCD00145430
InChI Key BWMISRWJRUSYEX-SZKNIZGXSA-N
Synonym terbinafine hydrochloride, terbinafine hcl, lamisil, terbinafine hydrochioride, lamisil krem, lamisil at, zabel, terbinafinum latin, terbinafina spanish, zimig
PubChem CID 5282481
ChEBI CHEBI:77614
IUPAC Name (E)-N,6,6-trimethyl-N-(naphthalen-1-ylmethyl)hept-2-en-4-yn-1-amine;hydrochloride
SMILES CC(C)(C)C#CC=CCN(C)CC1=CC=CC2=CC=CC=C21.Cl

Specifications

Melting Point 214°C
Color White
Quantity 1 g
Formula Weight 327.90
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material trans-N-(6,6-Dimethyl-2-hepten-4-ynyl)-N-methyl-1-naphthylmethylamine Hydrochloride
RTECSNumber QJ8600100
TSCA No