missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-m-Coumaric Acid 99.0+%, TCI America™
$35.41 - $96.94
Chemical Identifiers
| CAS | 14755-02-3 |
|---|---|
| Molecular Formula | C9H8O3 |
| Molecular Weight (g/mol) | 164.16 |
| MDL Number | MFCD00004390 |
| InChI Key | KKSDGJDHHZEWEP-SNAWJCMRSA-N |
| Synonym | 3-hydroxycinnamic acid, 3-coumaric acid, m-coumaric acid, m-hydroxycinnamic acid, 3-3-hydroxyphenyl acrylic acid, e-3-3-hydroxyphenyl acrylic acid, trans-3-hydroxycinnamic acid, trans-3-hydroxycinnamate, cinnamic acid, m-hydroxy, 2e-3-3-hydroxyphenyl prop-2-enoic acid |
| PubChem CID | 637541 |
| ChEBI | CHEBI:32357 |
| IUPAC Name | (E)-3-(3-hydroxyphenyl)prop-2-enoic acid |
| SMILES | C1=CC(=CC(=C1)O)C=CC(=O)O |
Chemical Identifiers
| 14755-02-3 | |
| 164.16 | |
| KKSDGJDHHZEWEP-SNAWJCMRSA-N | |
| 637541 | |
| (E)-3-(3-hydroxyphenyl)prop-2-enoic acid |
| C9H8O3 | |
| MFCD00004390 | |
| 3-hydroxycinnamic acid, 3-coumaric acid, m-coumaric acid, m-hydroxycinnamic acid, 3-3-hydroxyphenyl acrylic acid, e-3-3-hydroxyphenyl acrylic acid, trans-3-hydroxycinnamic acid, trans-3-hydroxycinnamate, cinnamic acid, m-hydroxy, 2e-3-3-hydroxyphenyl prop-2-enoic acid | |
| CHEBI:32357 | |
| C1=CC(=CC(=C1)O)C=CC(=O)O |
Specifications
| 14755-02-3 | |
| White | |
| MFCD00004390 | |
| 3-hydroxycinnamic acid, 3-coumaric acid, m-coumaric acid, m-hydroxycinnamic acid, 3-3-hydroxyphenyl acrylic acid, e-3-3-hydroxyphenyl acrylic acid, trans-3-hydroxycinnamic acid, trans-3-hydroxycinnamate, cinnamic acid, m-hydroxy, 2e-3-3-hydroxyphenyl prop-2-enoic acid | |
| C1=CC(=CC(=C1)O)C=CC(=O)O | |
| 164.16 | |
| CHEBI:32357 | |
| ≥99.0% (GC,T) | |
| trans-m-Coumaric Acid |
| 196°C | |
| C9H8O3 | |
| 5 g | |
| KKSDGJDHHZEWEP-SNAWJCMRSA-N | |
| (E)-3-(3-hydroxyphenyl)prop-2-enoic acid | |
| 637541 | |
| 164.16 | |
| Crystalline Powder |
Safety and Handling
TSCA : No