missing translation for 'onlineSavingsMsg'
Learn More

trans-m-Coumaric Acid 99.0+%, TCI America™

Catalog No. C06555G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C06555G 5 g
C065525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C06555G Supplier TCI America Supplier No. C06555G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 14755-02-3
Molecular Formula C9H8O3
Molecular Weight (g/mol) 164.16
MDL Number MFCD00004390
InChI Key KKSDGJDHHZEWEP-SNAWJCMRSA-N
Synonym 3-hydroxycinnamic acid, 3-coumaric acid, m-coumaric acid, m-hydroxycinnamic acid, 3-3-hydroxyphenyl acrylic acid, e-3-3-hydroxyphenyl acrylic acid, trans-3-hydroxycinnamic acid, trans-3-hydroxycinnamate, cinnamic acid, m-hydroxy, 2e-3-3-hydroxyphenyl prop-2-enoic acid
PubChem CID 637541
ChEBI CHEBI:32357
IUPAC Name (E)-3-(3-hydroxyphenyl)prop-2-enoic acid
SMILES C1=CC(=CC(=C1)O)C=CC(=O)O

Specifications

Melting Point 196°C
Color White
Quantity 5 g
Formula Weight 164.16
Percent Purity ≥99.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material trans-m-Coumaric Acid