missing translation for 'onlineSavingsMsg'
Learn More

trans-Decahydronaphthalene 98.0+%, TCI America™

Catalog No. D0007500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0007500ML 500 mL
D000725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D0007500ML Supplier TCI America Supplier No. D0007500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 493-02-7
Molecular Formula C10H18
Molecular Weight (g/mol) 138.25
MDL Number MFCD00004130,MFCD00064189,MFCD00064191
InChI Key NNBZCPXTIHJBJL-UHFFFAOYSA-N
Synonym decalin, decahydronaphthalene, cis-decahydronaphthalene, trans-decahydronaphthalene, cis-decalin, dekalin, naphthalene, decahydro, perhydronaphthalene, trans-decalin, naphthalane
PubChem CID 7044
ChEBI CHEBI:38853
IUPAC Name decahydronaphthalene
SMILES C1CCC2CCCCC2C1

Specifications

Color Colorless
Boiling Point 185°C
Quantity 500 mL
UN Number 1147
Formula Weight 138.25
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material trans-Decahydronaphthalene
EINECSNumber (4)-0575
RTECSNumber QJ3150000
TSCA Yes