missing translation for 'onlineSavingsMsg'
Learn More

trans-Crotonic acid, 98%

CAS: 107-93-7 | C4H6O2 | 86.09 g/mol

Supplier:  Thermo Scientific Chemicals A1576536

 View more versions of this product

Catalog No. AAA1576536


Only null left

Description

Description

trans-Crotonic acid is used in the copolymerization of crotonic acid hydrogel systems by using gamma-rays. The crosslinked copolymeric crotonic acid hydrogels release fertilizers and drugs that help to prevent environmental pollution.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-Crotonic acid is used in the copolymerization of crotonic acid hydrogel systems by using gamma-rays. The crosslinked copolymeric crotonic acid hydrogels release fertilizers and drugs that help to prevent environmental pollution.

Solubility
Soluble in water.

Notes
Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.
Specifications

Specifications

trans-Crotonic acid
70°C to 74°C
128°C (100 mmHg)
C4H6O2
500 g
1719943
crotonic acid, e-but-2-enoic acid, but-2-enoic acid, trans-2-butenoic acid, trans-crotonic acid, 3-methylacrylic acid, e-2-butenoic acid, 2-butenoic acid, 2e-but-2-enoic acid, e-crotonic acid
LDHQCZJRKDOVOX-NSCUHMNNSA-N
(E)-but-2-enoic acid
637090
86.09
107-93-7
1.027
87°C (188°F)
MFCD00002701
UN2823
14,2597
Soluble in water.
CC=CC(=O)O
86.09
CHEBI:41131
98%

Chemical Identifiers

107-93-7
86.09
LDHQCZJRKDOVOX-NSCUHMNNSA-N
637090
(E)-but-2-enoic acid
C4H6O2
MFCD00002701
crotonic acid, e-but-2-enoic acid, but-2-enoic acid, trans-2-butenoic acid, trans-crotonic acid, 3-methylacrylic acid, e-2-butenoic acid, 2-butenoic acid, 2e-but-2-enoic acid, e-crotonic acid
CHEBI:41131
CC=CC(=O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

P280-P305+P351+P338-P310

H318

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CROTONIC ACID, SOLID

EINECSNumber : 203-533-9

RTECSNumber : GQ2800000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only