missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-Crotonic acid, 98%
Description
trans-Crotonic acid is used in the copolymerization of crotonic acid hydrogel systems by using gamma-rays. The crosslinked copolymeric crotonic acid hydrogels release fertilizers and drugs that help to prevent environmental pollution.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
trans-Crotonic acid is used in the copolymerization of crotonic acid hydrogel systems by using gamma-rays. The crosslinked copolymeric crotonic acid hydrogels release fertilizers and drugs that help to prevent environmental pollution.
Solubility
Soluble in water.
Notes
Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.
trans-Crotonic acid is used in the copolymerization of crotonic acid hydrogel systems by using gamma-rays. The crosslinked copolymeric crotonic acid hydrogels release fertilizers and drugs that help to prevent environmental pollution.
Solubility
Soluble in water.
Notes
Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.
Specifications
Specifications
| Melting Point | 70°C to 74°C |
| Density | 1.027 |
| Boiling Point | 128°C (100 mmHg) |
| Flash Point | 87°C (188°F) |
| Quantity | 2500 g |
| UN Number | UN2823 |
| Beilstein | 1719943 |
| Merck Index | 14,2597 |
| Solubility Information | Soluble in water. |
| Formula Weight | 86.09 |
| Show More |
Safety and Handling
| Hazard Category | H318 |
| Hazard Statement | GHS H Statement H314-H318-H302 Causes severe skin burns and eye damage. Causes serious eye damage. Harmful if swallowed. |
| Precautionary Statement | P280-P305+P351+P338-P310 |
| DOTInformation | Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CROTONIC ACID, SOLID |
| EINECSNumber | 203-533-9 |
| RTECSNumber | GQ2800000 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only