missing translation for 'onlineSavingsMsg'
Learn More

Trans-Clopenthixol Hydrochloride, British Pharmacopoeia (BP) Reference Standard, MilliporeSigma™ Supelco™

Catalog No. 111022906
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022906 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111022906 Supplier Merck Emd Millipore Supplier No. BP561

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets, has been developed and issued under the Authority of the issuing Pharmacopoeia.

Specifications

CAS 58045-22-0
Quantity 25 mg
Molecular Formula C22H25ClN2OS · 2 HCl
Linear Formula C22H25ClN2OS · 2 HCl
Synonym trans-Clopenthixol dihydrochloride
Formula Weight 473.89
Grade BP