missing translation for 'onlineSavingsMsg'
Learn More

trans,cis-2,6-Nonadienal 95.0+%, TCI America™

Catalog No. p-7133990
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N08361G 1 g
N08365G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N08361G Supplier TCI America Supplier No. N08361G

Chemical Identifiers

CAS 557-48-2
Molecular Formula C9H14O
Molecular Weight (g/mol) 138.21
MDL Number MFCD00007009
InChI Key HZYHMHHBBBSGHB-ODYTWBPASA-N
Synonym trans-2,cis-6-nonadienal, violet leaf aldehyde, cucumber aldehyde, 2e,6z-nona-2,6-dienal, 2,6-nonadienal, 2e,6z, nona-2,6-dienal, e,z-2,6-nonadienal, 2e,6z-nonadienal, 2e,6z-nona-2,6-dien-1-al, 2-trans-6-cis-nonadienal
PubChem CID 643731
ChEBI CHEBI:7610
IUPAC Name (2E,6Z)-nona-2,6-dienal
SMILES CCC=CCCC=CC=O

Specifications

Color Yellow
Boiling Point 90°C
Quantity 1 g
Formula Weight 138.21
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material trans,cis-2,6-Nonadienal
RTECSNumber RA5391800
TSCA Yes