missing translation for 'onlineSavingsMsg'
Learn More

trans,cis-2,6-Nonadienal 95.0+%, TCI America™

Supplier:  TCI America N08365G

 View more versions of this product

Catalog No. N08365G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

trans,cis-2,6-Nonadienal
Yellow
C9H14O
5 g
HZYHMHHBBBSGHB-ODYTWBPASA-N
(2E,6Z)-nona-2,6-dienal
643731
138.21
Liquid
557-48-2
90°C
MFCD00007009
trans-2,cis-6-nonadienal, violet leaf aldehyde, cucumber aldehyde, 2e,6z-nona-2,6-dienal, 2,6-nonadienal, 2e,6z, nona-2,6-dienal, e,z-2,6-nonadienal, 2e,6z-nonadienal, 2e,6z-nona-2,6-dien-1-al, 2-trans-6-cis-nonadienal
CCC=CCCC=CC=O
138.21
CHEBI:7610
≥95.0% (GC)

Chemical Identifiers

557-48-2
138.21
HZYHMHHBBBSGHB-ODYTWBPASA-N
643731
(2E,6Z)-nona-2,6-dienal
C9H14O
MFCD00007009
trans-2,cis-6-nonadienal, violet leaf aldehyde, cucumber aldehyde, 2e,6z-nona-2,6-dienal, 2,6-nonadienal, 2e,6z, nona-2,6-dienal, e,z-2,6-nonadienal, 2e,6z-nonadienal, 2e,6z-nona-2,6-dien-1-al, 2-trans-6-cis-nonadienal
CHEBI:7610
CCC=CCCC=CC=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : RA5391800

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions