missing translation for 'onlineSavingsMsg'
Learn More

trans-Cinnamic Acid 98.0+%, TCI America™

Catalog No. p-7124366
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C035325G 25 g
C0353100G 100 g
C0353500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C035325G Supplier TCI America Supplier No. C035325G
Only null left

Chemical Identifiers

CAS 140-10-3
Molecular Formula C9H8O2
Molecular Weight (g/mol) 148.161
MDL Number MFCD00004369
InChI Key WBYWAXJHAXSJNI-VOTSOKGWSA-N
Synonym cinnamic acid, trans-cinnamic acid, e-cinnamic acid, 3-phenylacrylic acid, trans-3-phenylacrylic acid, zimtsaeure, 2e-3-phenylprop-2-enoic acid, 3-phenylprop-2-enoic acid, 3-phenylpropenoic acid
PubChem CID 444539
ChEBI CHEBI:35697
IUPAC Name (E)-3-phenylprop-2-enoic acid
SMILES C1=CC=C(C=C1)C=CC(=O)O

Specifications

Melting Point 135°C
Color White-Yellow
Boiling Point 300°C
Quantity 25 g
Formula Weight 148.16
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material trans-Cinnamic Acid
EINECSNumber (3)-1719
RTECSNumber GD7850000
TSCA Yes