missing translation for 'onlineSavingsMsg'
Learn More

trans-Cinnamic Acid, Analytical Standard, ≥98.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111013049
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013049 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013049 Supplier Merck Emd Millipore Supplier No. 9701350MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

trans-Cinnamic acid occurs in plants, formed via deamination of L-phenylalanine in the presence of enzymatic catalyst, L-phenylalanine ammonia-lyase.

Specifications

CAS 140-10-3
Melting Point 133°C; 133°C to 137°C
Packaging Bottomless Glass Bottle with Fused Cone
Linear Formula C9H8O2
MDL Number MFCD00004369
Quantity 50 mg
Synonym (2 E)-3-Phenyl-2-propenoic acid; trans-3-Phenylacrylic acid
Formula Weight 148.16
Grade Analytical Standard
Physical Form Neat