missing translation for 'onlineSavingsMsg'
Learn More

trans-Cinnamamide 98.0+%, TCI America™

Catalog No. p-7124490
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C05365G 5 g
C053625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C05365G Supplier TCI America Supplier No. C05365G
Only null left

Chemical Identifiers

CAS 22031-64-7
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.177
MDL Number MFCD00008033
InChI Key APEJMQOBVMLION-VOTSOKGWSA-N
Synonym trans-3-Phenylacrylamide
PubChem CID 5273472
ChEBI CHEBI:76320
IUPAC Name (E)-3-phenylprop-2-enamide
SMILES C1=CC=C(C=C1)C=CC(=O)N

Specifications

Melting Point 150°C
Color White-Yellow
Quantity 5 g
Formula Weight 147.18
Percent Purity ≥98.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material trans-Cinnamamide
RTECSNumber GD6853000
TSCA No