missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-Cinnamaldehyde 98.0+%, TCI America™
Supplier: TCI America C035225ML
Specifications
| trans-Cinnamaldehyde | |
| -8°C | |
| 252°C | |
| MFCD00007000 | |
| cinnamaldehyde, trans-cinnamaldehyde, cinnamic aldehyde, e-cinnamaldehyde, cinnamal, 3-phenylacrylaldehyde, zimtaldehyde, cinnamylaldehyde, 3-phenylpropenal, 2e-3-phenylprop-2-enal | |
| O=C\C=C\C1=CC=CC=C1 | |
| 132.16 | |
| CHEBI:16731 | |
| ≥98.0% (GC) |
| 14371-10-9 | |
| Yellow | |
| C9H8O | |
| 25 mL | |
| KJPRLNWUNMBNBZ-QPJJXVBHSA-N | |
| (2E)-3-phenylprop-2-enal | |
| 637511 | |
| 132.16 | |
| Liquid |
Chemical Identifiers
| 14371-10-9 | |
| 132.16 | |
| KJPRLNWUNMBNBZ-QPJJXVBHSA-N | |
| 637511 | |
| (2E)-3-phenylprop-2-enal |
| C9H8O | |
| MFCD00007000 | |
| cinnamaldehyde, trans-cinnamaldehyde, cinnamic aldehyde, e-cinnamaldehyde, cinnamal, 3-phenylacrylaldehyde, zimtaldehyde, cinnamylaldehyde, 3-phenylpropenal, 2e-3-phenylprop-2-enal | |
| CHEBI:16731 | |
| O=C\C=C\C1=CC=CC=C1 |
Safety and Handling
EINECSNumber : (3)-1148
RTECSNumber : GD6476000
TSCA : Yes