missing translation for 'onlineSavingsMsg'
Learn More

trans-Cinnamaldehyde, 98%, Spectrum™ Chemical

Catalog No. p-7165437
Change view
Click to view available options
Quantity:
1 kg
200 L
Packaging:
Amber Glass Bottle
Drum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18614497 200 L Drum
18614496 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18614497 Supplier Spectrum Chemical Mfg Cor Supplier No. CI110200LTBL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 14371-10-9

Specifications

CAS Max % 1
Packaging Drum
Quantity 200 L
Percent Purity 0.98
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.