Learn More
trans-Bis(benzonitrile)dichloropalladium(II), Pd 27.1%, Thermo Scientific Chemicals
$293.04 - $293.04
Chemical Identifiers
| CAS | 14220-64-5 |
|---|---|
| Molecular Formula | C14H10Cl2N2Pd |
| Molecular Weight (g/mol) | 383.57 |
| MDL Number | MFCD00013123 |
| InChI Key | QQPSWGUJSPTOSH-UHFFFAOYSA-L |
| Synonym | bis benzonitrile palladium chloride, bis benzonitrile palladium ii chloride, trans-bis benzonitrile palladium ii chloride, bis benzonitrile palladium ii chloride, 500mg, palladium 2+ bis benzonitrile dichloride |
| PubChem CID | 10271322 |
| IUPAC Name | benzonitrile;palladium(2+);dichloride |
| SMILES | Cl[Pd++](Cl)([N]#CC1=CC=CC=C1)[N]#CC1=CC=CC=C1 |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 14220-64-5 | |
| 383.57 | |
| QQPSWGUJSPTOSH-UHFFFAOYSA-L | |
| 10271322 | |
| Cl[Pd++](Cl)([N]#CC1=CC=CC=C1)[N]#CC1=CC=CC=C1 |
| C14H10Cl2N2Pd | |
| MFCD00013123 | |
| bis benzonitrile palladium chloride, bis benzonitrile palladium ii chloride, trans-bis benzonitrile palladium ii chloride, bis benzonitrile palladium ii chloride, 500mg, palladium 2+ bis benzonitrile dichloride | |
| benzonitrile;palladium(2+);dichloride |
Specifications
| 14220-64-5 | |
| C14H10Cl2N2Pd | |
| MFCD00013123 | |
| UN3439 | |
| Soluble in acetone, chloroform | |
| Cl[Pd++](Cl)([N]#CC1=CC=CC=C1)[N]#CC1=CC=CC=C1 | |
| 383.57 | |
| 383.57 | |
| Powder |
| White | |
| PdCl2[(C6H5)(CN)]2 | |
| 1 g | |
| bis benzonitrile palladium chloride, bis benzonitrile palladium ii chloride, trans-bis benzonitrile palladium ii chloride, bis benzonitrile palladium ii chloride, 500mg, palladium 2+ bis benzonitrile dichloride | |
| QQPSWGUJSPTOSH-UHFFFAOYSA-L | |
| benzonitrile;palladium(2+);dichloride | |
| 10271322 | |
| Pd 27.1% | |
| trans-Bis(benzonitrile)dichloropalladium(II) |
Safety and Handling
GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P501c
H301+H311+H331-H315-H319
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber : 238-085-3
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only