missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-Aconitic Acid 98.0+%, TCI America™
$119.30 - $1112.84
Chemical Identifiers
| CAS | 4023-65-8 |
|---|---|
| Molecular Formula | C6H6O6 |
| Molecular Weight (g/mol) | 174.108 |
| MDL Number | MFCD00002721 |
| InChI Key | GTZCVFVGUGFEME-HNQUOIGGSA-N |
| Synonym | trans-aconitic acid, trans-aconitate, e-1-propene-1,2,3-tricarboxylic acid, aconitic acid, e-prop-1-ene-1,2,3-tricarboxylic acid, aconitic acid, trans, trans aconitic acid, e-aconitic acid, unii-7db37960cw, 1e-1-propene-1,2,3-tricarboxylic acid |
| PubChem CID | 444212 |
| ChEBI | CHEBI:32806 |
| IUPAC Name | (E)-prop-1-ene-1,2,3-tricarboxylic acid |
| SMILES | C(C(=CC(=O)O)C(=O)O)C(=O)O |
Chemical Identifiers
| 4023-65-8 | |
| 174.108 | |
| GTZCVFVGUGFEME-HNQUOIGGSA-N | |
| 444212 | |
| (E)-prop-1-ene-1,2,3-tricarboxylic acid |
| C6H6O6 | |
| MFCD00002721 | |
| trans-aconitic acid, trans-aconitate, e-1-propene-1,2,3-tricarboxylic acid, aconitic acid, e-prop-1-ene-1,2,3-tricarboxylic acid, aconitic acid, trans, trans aconitic acid, e-aconitic acid, unii-7db37960cw, 1e-1-propene-1,2,3-tricarboxylic acid | |
| CHEBI:32806 | |
| C(C(=CC(=O)O)C(=O)O)C(=O)O |
Specifications
| 4023-65-8 | |
| White-Yellow | |
| MFCD00002721 | |
| trans-aconitic acid, trans-aconitate, e-1-propene-1,2,3-tricarboxylic acid, aconitic acid, e-prop-1-ene-1,2,3-tricarboxylic acid, aconitic acid, trans, trans aconitic acid, e-aconitic acid, unii-7db37960cw, 1e-1-propene-1,2,3-tricarboxylic acid | |
| C(C(=CC(=O)O)C(=O)O)C(=O)O | |
| 174.108 | |
| CHEBI:32806 | |
| ≥98.0% (HPLC,T) | |
| trans-Aconitic Acid |
| 180°C | |
| C6H6O6 | |
| 25 g | |
| GTZCVFVGUGFEME-HNQUOIGGSA-N | |
| (E)-prop-1-ene-1,2,3-tricarboxylic acid | |
| 444212 | |
| 174.11 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : UD2380000
TSCA : Yes