missing translation for 'onlineSavingsMsg'
Learn More

trans-Aconitic Acid 98.0+%, TCI America™

Catalog No. A012725G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A012725G 25 g
A0127500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A012725G Supplier TCI America Supplier No. A012725G

Chemical Identifiers

CAS 4023-65-8
Molecular Formula C6H6O6
Molecular Weight (g/mol) 174.108
MDL Number MFCD00002721
InChI Key GTZCVFVGUGFEME-HNQUOIGGSA-N
Synonym trans-aconitic acid, trans-aconitate, e-1-propene-1,2,3-tricarboxylic acid, aconitic acid, e-prop-1-ene-1,2,3-tricarboxylic acid, aconitic acid, trans, trans aconitic acid, e-aconitic acid, unii-7db37960cw, 1e-1-propene-1,2,3-tricarboxylic acid
PubChem CID 444212
ChEBI CHEBI:32806
IUPAC Name (E)-prop-1-ene-1,2,3-tricarboxylic acid
SMILES C(C(=CC(=O)O)C(=O)O)C(=O)O

Specifications

Melting Point 180°C
Color White-Yellow
Quantity 25 g
Formula Weight 174.11
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material trans-Aconitic Acid
RTECSNumber UD2380000
TSCA Yes