missing translation for 'onlineSavingsMsg'
Learn More

trans-Aconitic acid, 98%

CAS: 4023-65-8 | C6H6O6 | 174.108 g/mol

$109.81 - $356.59

Chemical Identifiers

CAS 4023-65-8
Molecular Formula C6H6O6
Molecular Weight (g/mol) 174.108
MDL Number MFCD00002721
InChI Key GTZCVFVGUGFEME-HNQUOIGGSA-N
Synonym trans-aconitic acid, trans-aconitate, e-1-propene-1,2,3-tricarboxylic acid, aconitic acid, e-prop-1-ene-1,2,3-tricarboxylic acid, aconitic acid, trans, trans aconitic acid, e-aconitic acid, unii-7db37960cw, 1e-1-propene-1,2,3-tricarboxylic acid
PubChem CID 444212
ChEBI CHEBI:32806
IUPAC Name (E)-prop-1-ene-1,2,3-tricarboxylic acid
SMILES C(C(=CC(=O)O)C(=O)O)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2008714
View Documents
Thermo Scientific Chemicals
B2008714
25 g
Each for $109.81
Only null left
 
AAB2008722
View Documents
Thermo Scientific Chemicals
B2008722
100 g
Each for $356.59
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
It plays an important role as an antimicrobial agent that blocks the transformation of Leishmania donovani amastigotes to prostigotes. trans-Aconitic acid is also used in synthesis of aconitate esters. It may be used for cross-linking polyvinyl alcohol film for preparing composite nanofiltration membranes.

Solubility
Soluble in water at 20°C 400g/L. Slightly soluble in ether.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Chemical Identifiers

4023-65-8
174.108
GTZCVFVGUGFEME-HNQUOIGGSA-N
444212
(E)-prop-1-ene-1,2,3-tricarboxylic acid
C6H6O6
MFCD00002721
trans-aconitic acid, trans-aconitate, e-1-propene-1,2,3-tricarboxylic acid, aconitic acid, e-prop-1-ene-1,2,3-tricarboxylic acid, aconitic acid, trans, trans aconitic acid, e-aconitic acid, unii-7db37960cw, 1e-1-propene-1,2,3-tricarboxylic acid
CHEBI:32806
C(C(=CC(=O)O)C(=O)O)C(=O)O

Specifications

4023-65-8
Odorless
MFCD00002721
14,117
Soluble in water at 20°C 400g/L. Slightly soluble in ether.
C(C(=CC(=O)O)C(=O)O)C(=O)O
174.108
CHEBI:32806
98%
∼190°C (decomposition)
C6H6O6
25 g
trans-aconitic acid, trans-aconitate, e-1-propene-1,2,3-tricarboxylic acid, aconitic acid, e-prop-1-ene-1,2,3-tricarboxylic acid, aconitic acid, trans, trans aconitic acid, e-aconitic acid, unii-7db37960cw, 1e-1-propene-1,2,3-tricarboxylic acid
GTZCVFVGUGFEME-HNQUOIGGSA-N
(E)-prop-1-ene-1,2,3-tricarboxylic acid
444212
174.11
trans-Aconitic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 223-688-6

RTECSNumber : UD2380000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only