missing translation for 'onlineSavingsMsg'
Learn More

trans-9-Octadecenol 99.0+%, TCI America™

Catalog No. O02281G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O02281G 1 g
O02285G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O02281G Supplier TCI America Supplier No. O02281G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 506-42-3
Molecular Formula C18H36O
Molecular Weight (g/mol) 268.485
MDL Number MFCD00063218
InChI Key ALSTYHKOOCGGFT-MDZDMXLPSA-N
Synonym Elaidyl Alcohol
PubChem CID 5367665
IUPAC Name (E)-octadec-9-en-1-ol
SMILES CCCCCCCCC=CCCCCCCCCO

Specifications

Melting Point 37°C
Color White
Boiling Point 333°C
Quantity 1 g
Formula Weight 268.49
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material trans-9-Octadecenol
EINECSNumber (2)-0258
TSCA No
Recommended Storage Refrigerator