missing translation for 'onlineSavingsMsg'
Learn More

trans-5-Decene, 97%

Catalog No. AAL1068906
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1068906 5 g
AAL1068903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1068906 Supplier Thermo Scientific Chemicals Supplier No. L1068906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7433-56-9 | C10H20 | 140.27 g/mol

trans-5-Decene has been used to characterize the effect of cyclization and size on the stability of Criegee intermediates formed in ozonolysis. It has been added to phenoxathiin cation radical in acetonitrile solution to form bis(10-phenoxathiiniumyl) alkane adducts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-5-Decene has been used to characterize the effect of cyclization and size on the stability of Criegee intermediates formed in ozonolysis. It has been added to phenoxathiin cation radical in acetonitrile solution to form bis(10-phenoxathiiniumyl) alkane adducts.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 7433-56-9
Molecular Formula C10H20
Molecular Weight (g/mol) 140.27
MDL Number MFCD00009508
InChI Key UURSXESKOOOTOV-MDZDMXLPSA-N
Synonym trans-5-decene, e-5-decene, 5-decene, e, trans-dec-5-ene, 5e-decene, 5-decene, cis-5-decene, 5-decene, 5e, 5-trans-decene, 5e dec-5-ene
PubChem CID 637820
IUPAC Name (E)-dec-5-ene
SMILES CCCC\C=C\CCCC

Specifications

Density 0.74
Boiling Point 171°C to 172°C
Flash Point 46°C (114°F)
Refractive Index 1.424
Quantity 5 g
UN Number UN3295
Beilstein 1719487
Solubility Information Insoluble in water.
Formula Weight 140.27
Percent Purity 97%
Chemical Name or Material trans-5-Decene
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
EINECSNumber 231-080-7
RTECSNumber HE2080000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only