missing translation for 'onlineSavingsMsg'
Learn More

trans-5-Decen-1-ol, 96%

Catalog No. AAL1176803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1176803 1 g
AAL1176806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1176803 Supplier Thermo Scientific Chemicals Supplier No. L1176803
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 56578-18-8 | C10H20O | 156.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 56578-18-8
Molecular Formula C10H20O
Molecular Weight (g/mol) 156.27
MDL Number MFCD00056187
InChI Key WYPQHXVMNVEVEB-AATRIKPKSA-N
Synonym trans-5-decen-1-ol, e-5-decen-1-ol, trans-5-decenol, 5-decen-1-ol, 5e, 5-decen-1-ol, e, decan-5-en-1-ol, e-5-decenol, unii-m709v74ozv, 5-decen-1-ol, epa pesticide chemical code 078038
PubChem CID 5283292
IUPAC Name (E)-dec-5-en-1-ol
SMILES CCCC\C=C\CCCCO

Specifications

Boiling Point 113°C to 118°C (11 mmHg)
Flash Point 62°C (143°F)
Refractive Index 1.452
Quantity 1 g
Beilstein 1840630
Formula Weight 156.27
Percent Purity 96%
Chemical Name or Material trans-5-Decen-1-ol
Hazard Category H227
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 260-267-6
RTECSNumber HE2096000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only