Learn More
trans-4-(Trifluoromethyl)cinnamic acid, 98%
CAS: 16642-92-5 | C10H6F3O2 | 215.15 g/mol
Supplier: Thermo Scientific Chemicals L0277106
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| trans-4-(Trifluoromethyl)cinnamic acid | |
| 230°C to 234°C | |
| C10H6F3O2 | |
| 5 g | |
| 4-trifluoromethyl cinnamic acid, trans-4-trifluoromethyl cinnamic acid, p-trifluoromethyl cinnamic acid, e-3-4-trifluoromethyl phenyl acrylic acid, 2e-3-4-trifluoromethyl phenyl prop-2-enoic acid, 4-trifluoromethylcinnamic acid, 2-propenoic acid, 3-4-trifluoromethyl phenyl, 3-4-trifluoromethyl-phenyl-acrylic acid, ptf-cna, trans-4-trifluoromethylcinnamic acid | |
| [O-]C(=O)\C=C\C1=CC=C(C=C1)C(F)(F)F | |
| 215.15 | |
| CHEBI:60705 | |
| 98% |
| 16642-92-5 | |
| Odorless | |
| MFCD00002696 | |
| 2452800 | |
| ANRMAUMHJREENI-ZZXKWVIFSA-M | |
| (E)-3-[4-(trifluoromethyl)phenyl]prop-2-enoic acid | |
| 688070 | |
| 216.16 |
Chemical Identifiers
| 16642-92-5 | |
| 215.15 | |
| ANRMAUMHJREENI-ZZXKWVIFSA-M | |
| 688070 | |
| (E)-3-[4-(trifluoromethyl)phenyl]prop-2-enoic acid |
| C10H6F3O2 | |
| MFCD00002696 | |
| 4-trifluoromethyl cinnamic acid, trans-4-trifluoromethyl cinnamic acid, p-trifluoromethyl cinnamic acid, e-3-4-trifluoromethyl phenyl acrylic acid, 2e-3-4-trifluoromethyl phenyl prop-2-enoic acid, 4-trifluoromethylcinnamic acid, 2-propenoic acid, 3-4-trifluoromethyl phenyl, 3-4-trifluoromethyl-phenyl-acrylic acid, ptf-cna, trans-4-trifluoromethylcinnamic acid | |
| CHEBI:60705 | |
| [O-]C(=O)\C=C\C1=CC=C(C=C1)C(F)(F)F |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only