missing translation for 'onlineSavingsMsg'
Learn More

trans-4-(Trifluoromethyl)cinnamic acid, 98%

Catalog No. AAL0277103
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0277103 1 g
AAL0277106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0277103 Supplier Thermo Scientific Chemicals Supplier No. L0277103
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 16642-92-5 | C10H6F3O2 | 215.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16642-92-5
Molecular Formula C10H6F3O2
Molecular Weight (g/mol) 215.15
MDL Number MFCD00002696
InChI Key ANRMAUMHJREENI-ZZXKWVIFSA-M
Synonym 4-trifluoromethyl cinnamic acid, trans-4-trifluoromethyl cinnamic acid, p-trifluoromethyl cinnamic acid, e-3-4-trifluoromethyl phenyl acrylic acid, 2e-3-4-trifluoromethyl phenyl prop-2-enoic acid, 4-trifluoromethylcinnamic acid, 2-propenoic acid, 3-4-trifluoromethyl phenyl, 3-4-trifluoromethyl-phenyl-acrylic acid, ptf-cna, trans-4-trifluoromethylcinnamic acid
PubChem CID 688070
ChEBI CHEBI:60705
IUPAC Name (E)-3-[4-(trifluoromethyl)phenyl]prop-2-enoic acid
SMILES [O-]C(=O)\C=C\C1=CC=C(C=C1)C(F)(F)F

Specifications

Melting Point 230°C to 234°C
Odor Odorless
Quantity 1 g
Beilstein 2452800
Formula Weight 216.16
Percent Purity 98%
Chemical Name or Material trans-4-(Trifluoromethyl)cinnamic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only