missing translation for 'onlineSavingsMsg'
Learn More

trans-4-(tert-Butyldiphenylsilyloxy)-L-proline 98.0+%, TCI America™

Catalog No. B34401G
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Catalog No. Quantity
B34401G 1 g
B34405G 5 g
2 options

Catalog No. B34401G

Supplier: TCI America B34401G

Only null left

Chemical Identifiers

CAS 259212-61-8
Molecular Formula C21H27NO3Si
Molecular Weight (g/mol) 369.54
MDL Number MFCD12407126
InChI Key LHSRURYOCDIZEW-UHFFFAOYNA-N
Synonym (2S,4R)-4-(tert-Butyldiphenylsilyloxy)pyrrolidine-2-carboxylic Acid
PubChem CID 11552425
IUPAC Name 4-[(tert-butyldiphenylsilyl)oxy]pyrrolidine-2-carboxylic acid
SMILES CC(C)(C)[Si](OC1CNC(C1)C(O)=O)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 198°C
Color White
Quantity 1 g
Formula Weight 369.54
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material trans-4-(tert-Butyldiphenylsilyloxy)-L-proline