missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-4-(tert-Butoxycarbonylaminomethyl)cyclohexanecarboxylic Acid 98.0+%, TCI America™
$251.65 - $885.43
Chemical Identifiers
| CAS | 27687-14-5 |
|---|---|
| Molecular Formula | C13H23NO4 |
| Molecular Weight (g/mol) | 257.33 |
| InChI Key | AZEKNJGFCSHZID-UHFFFAOYSA-N |
| Synonym | trans-4-(Boc-aminomethyl)cyclohexanecarboxylic Acid |
| PubChem CID | 736918 |
| IUPAC Name | 4-[[(2-methylpropan-2-yl)oxycarbonylamino]methyl]cyclohexane-1-carboxylic acid |
| SMILES | CC(C)(C)OC(=O)NCC1CCC(CC1)C(=O)O |
Chemical Identifiers
| 27687-14-5 | |
| 257.33 | |
| trans-4-(Boc-aminomethyl)cyclohexanecarboxylic Acid | |
| 4-[[(2-methylpropan-2-yl)oxycarbonylamino]methyl]cyclohexane-1-carboxylic acid |
| C13H23NO4 | |
| AZEKNJGFCSHZID-UHFFFAOYSA-N | |
| 736918 | |
| CC(C)(C)OC(=O)NCC1CCC(CC1)C(=O)O |
Specifications
| 135°C | |
| White-Yellow | |
| trans-4-(Boc-aminomethyl)cyclohexanecarboxylic Acid | |
| CC(C)(C)OC(=O)NCC1CCC(CC1)C(=O)O | |
| 5 g | |
| 736918 | |
| ≥98.0% (GC,T) | |
| trans-4-(tert-Butoxycarbonylaminomethyl)cyclohexanecarboxylic Acid |
| 27687-14-5 | |
| C13H23NO4 | |
| AZEKNJGFCSHZID-UHFFFAOYSA-N | |
| 4-[[(2-methylpropan-2-yl)oxycarbonylamino]methyl]cyclohexane-1-carboxylic acid | |
| 257.33 | |
| 257.33 | |
| Crystalline Powder |
Safety and Handling
TSCA : No