missing translation for 'onlineSavingsMsg'
Learn More

trans-4-(tert-Butoxycarbonylamino)cyclohexanol 98.0+%, TCI America™

Catalog No. B35645G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B35645G 5 g
B35641G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B35645G Supplier TCI America Supplier No. B35645G

Chemical Identifiers

CAS 111300-06-2
Molecular Formula C11H21NO3
Molecular Weight (g/mol) 215.29
MDL Number MFCD03844613,MFCD03844614,MFCD06658349
InChI Key DQARDWKWPIRJEH-UHFFFAOYSA-N
Synonym trans-4-boc-aminocyclohexanol, tert-butyl 4-hydroxycyclohexyl carbamate, boc-trans-4-aminocyclohexanol, 4-boc-amino cyclohexanol, tert-butyl trans-4-hydroxycyclohexyl carbamate, tert-butyl n-4-hydroxycyclohexyl carbamate, tert-butyl cis-4-hydroxycyclohexylcarbamate, trans-n-boc-4-amino-cyclohexanol, 4-n-boc-amino-cyclohexanol, tert-butyl cis-4-hydroxycyclohexyl carbamate
PubChem CID 1514287
IUPAC Name tert-butyl N-(4-hydroxycyclohexyl)carbamate
SMILES CC(C)(C)OC(=O)NC1CCC(O)CC1

Specifications

Melting Point 171°C
Color White-Yellow
Quantity 5 g
Formula Weight 215.29
Percent Purity ≥98.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material trans-4-(tert-Butoxycarbonylamino)cyclohexanol