missing translation for 'onlineSavingsMsg'
Learn More

trans-4-Pentylcyclohexanecarboxylic Acid 99.0+%, TCI America™

Catalog No. p-7134863
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P09445G 5 g
P094425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P09445G Supplier TCI America Supplier No. P09445G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 38289-29-1
Molecular Formula C12H22O2
Molecular Weight (g/mol) 198.306
MDL Number MFCD00009964
InChI Key RVLAXPQGTRTHEV-UHFFFAOYSA-N
Synonym trans-4-pentylcyclohexanecarboxylic acid, 4-pentylcyclohexanecarboxylic acid, trans-4-n-pentylcyclohexanecarboxylic acid, cyclohexanecarboxylic acid, 4-pentyl, trans-4-amylcyclohexanecarboxylic acid, cyclohexanecarboxylic acid, 4-pentyl-, trans, 4-n-pentylcyclohexane carboxylic acid, 4-amylcyclohexanecarboxylic acid, rvlaxpqgtrthev-xypyzodxsa-n, 1s,4r-4-pentylcyclohexane-1-carboxylic acid
PubChem CID 98882
IUPAC Name 4-pentylcyclohexane-1-carboxylic acid
SMILES CCCCCC1CCC(CC1)C(=O)O

Specifications

Melting Point 55°C
Color White
Quantity 5 g
Formula Weight 198.31
Percent Purity ≥99.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material trans-4-Pentylcyclohexanecarboxylic Acid
EINECSNumber (3)-3723
TSCA No