missing translation for 'onlineSavingsMsg'
Learn More

trans-4-Octene 99.0+%, TCI America™

Catalog No. p-7134202
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O01361ML 1 mL
O01365ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O01361ML Supplier TCI America Supplier No. O01361ML

Chemical Identifiers

CAS 14850-23-8
Molecular Formula C8H16
Molecular Weight (g/mol) 112.216
MDL Number MFCD00009476
InChI Key IRUCBBFNLDIMIK-BQYQJAHWSA-N
Synonym trans-4-octene, e-4-octene, trans-n-4-octene, e-oct-4-ene, 4-octene, e, 4-octene, 4-octene, 4e, unii-fkz5737b1w, n-trans-4-octene, 4e-4-octene
PubChem CID 5357253
IUPAC Name (E)-oct-4-ene
SMILES CCCC=CCCC

Specifications

Color Colorless
Boiling Point 122°C
Quantity 1 mL
UN Number 3295
Formula Weight 112.22
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material trans-4-Octene
EINECSNumber (2)-0024
TSCA No