missing translation for 'onlineSavingsMsg'
Learn More

trans-4-Octene, 97%

Catalog No. AA4315504
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4315504 2 g
AA4315509 10 g
AA4315518 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4315504 Supplier Thermo Scientific Chemicals Supplier No. 04315504

CAS: 14850-23-8 | C8H16 | 112.216 g/mol

It is used in pharmaceutical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14850-23-8
Molecular Formula C8H16
Molecular Weight (g/mol) 112.216
MDL Number MFCD00009476
InChI Key IRUCBBFNLDIMIK-BQYQJAHWSA-N
Synonym trans-4-octene, e-4-octene, trans-n-4-octene, e-oct-4-ene, 4-octene, e, 4-octene, 4-octene, 4e, unii-fkz5737b1w, n-trans-4-octene, 4e-4-octene
PubChem CID 5357253
IUPAC Name (E)-oct-4-ene
SMILES CCCC=CCCC

Specifications

Density 0.715 g/mL
Boiling Point 122°C to 123°C
Flash Point 21°C (70°F)
Odor Characteristic
Refractive Index 1.412
Quantity 2 g
UN Number UN3295
Beilstein 1719104
Solubility Information Sparingly soluble in water(10.7,mg/L).
Formula Weight 112.22
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material trans-4-Octene
Show More Show Less
Hazard Category H225
Hazard Statement GHS H Statement
H225
Highly flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
EINECSNumber 238-913-3
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only