missing translation for 'onlineSavingsMsg'
Learn More

trans-4-Methylcyclohexanol 98.0+%, TCI America™

Catalog No. M03545ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M03545ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M03545ML Supplier TCI America Supplier No. M03545ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7731-29-5
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.188
MDL Number MFCD00064171
InChI Key MQWCXKGKQLNYQG-UHFFFAOYSA-N
Synonym 4-methylcyclohexanol, trans-4-methylcyclohexanol, cis-4-methylcyclohexanol, cyclohexanol, 4-methyl, 1-methyl-4-cyclohexanol, cyclohexanol, 4-methyl-, trans, methyl cyclohexanol, p-methylcyclohexanol, cyclohexanol, 4-methyl-, cis, trans 4-methyl cyclohexanol
PubChem CID 11524
IUPAC Name 4-methylcyclohexan-1-ol
SMILES CC1CCC(CC1)O

Specifications

Color Colorless
Boiling Point 171°C
Quantity 5 mL
Formula Weight 114.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material trans-4-Methylcyclohexanol
EINECSNumber (3)-2301
TSCA No