missing translation for 'onlineSavingsMsg'
Learn More

trans-4-Methylcyclohexanol, 98%

Catalog No. p-7024825
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2005406 5 g
AAB2005414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2005406 Supplier Thermo Scientific Chemicals Supplier No. B2005406
Only null left

CAS: 7731-29-5 | C7H14O | 114.188 g/mol

trans-4-Methylcyclohexanol is used to prepare 4-methylcyclohexyl nitrite to study the laser-induced fluorescence (LIF) excitation spectra of the 4-methylcyclohexoxy and d11-cyclohexoxy radicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-4-Methylcyclohexanol is used to prepare 4-methylcyclohexyl nitrite to study the laser-induced fluorescence (LIF) excitation spectra of the 4-methylcyclohexoxy and d11-cyclohexoxy radicals.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 7731-29-5
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.188
MDL Number MFCD00064171
InChI Key MQWCXKGKQLNYQG-UHFFFAOYSA-N
Synonym 4-methylcyclohexanol, trans-4-methylcyclohexanol, cis-4-methylcyclohexanol, cyclohexanol, 4-methyl, 1-methyl-4-cyclohexanol, cyclohexanol, 4-methyl-, trans, methyl cyclohexanol, p-methylcyclohexanol, cyclohexanol, 4-methyl-, cis, trans 4-methyl cyclohexanol
PubChem CID 11524
IUPAC Name 4-methylcyclohexan-1-ol
SMILES CC1CCC(CC1)O

Specifications

Density 0.91
Boiling Point 173°C to 175°C
Flash Point 72°C (162°F)
Refractive Index 1.455
Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 114.19
Percent Purity 98%
Chemical Name or Material trans-4-Methylcyclohexanol
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 231-790-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only