missing translation for 'onlineSavingsMsg'
Learn More

trans-4-Methylcyclohexanecarboxylic Acid 98.0+%, TCI America™

Catalog No. p-7132854
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M17835G 5 g
M178325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M17835G Supplier TCI America Supplier No. M17835G

Chemical Identifiers

CAS 13064-83-0
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.20
MDL Number MFCD00074943,MFCD00074909,MFCD20617670
InChI Key QTDXSEZXAPHVBI-UHFFFAOYSA-N
Synonym trans-4-methylcyclohexanecarboxylic acid, 4-methylcyclohexanecarboxylic acid, 4-methyl-1-cyclohexanecarboxylic acid, cis-4-methylcyclohexanecarboxylic acid, unii-70f0i2amtv, unii-m7dsk28350, trans-4-methyl-1-cyclohexanecarboxylic acid, trans-4-methyl hexahydrobenzoic acid, trans-4-methylcyclohexane carboxylic acid, 70f0i2amtv
PubChem CID 20330
IUPAC Name 4-methylcyclohexane-1-carboxylic acid
SMILES CC1CCC(CC1)C(O)=O

Specifications

Melting Point 112°C
Color White-Yellow
Boiling Point 245°C
Quantity 5 g
Formula Weight 142.20
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material trans-4-Methylcyclohexanecarboxylic Acid
EINECSNumber (3)-3311
TSCA No