missing translation for 'onlineSavingsMsg'
Learn More

trans-4-Methylcyclohexanecarboxylic acid, 98%

Catalog No. p-7029933
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
AAH2745503 1 g
AAH2745506 5 g
2 options

Catalog No. AAH2745503

Supplier: Thermo Scientific Chemicals H2745503

Only null left

CAS: 13064-83-0 | C8H14O2 | 142.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13064-83-0
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.20
MDL Number MFCD00074943,MFCD00074909,MFCD20617670
InChI Key QTDXSEZXAPHVBI-UHFFFAOYSA-N
Synonym trans-4-methylcyclohexanecarboxylic acid, 4-methylcyclohexanecarboxylic acid, 4-methyl-1-cyclohexanecarboxylic acid, cis-4-methylcyclohexanecarboxylic acid, unii-70f0i2amtv, unii-m7dsk28350, trans-4-methyl-1-cyclohexanecarboxylic acid, trans-4-methyl hexahydrobenzoic acid, trans-4-methylcyclohexane carboxylic acid, 70f0i2amtv
PubChem CID 20330
IUPAC Name 4-methylcyclohexane-1-carboxylic acid
SMILES CC1CCC(CC1)C(O)=O

Specifications

Melting Point 109°C to 110°C
Quantity 1 g
Formula Weight 142.2
Percent Purity 98%
Chemical Name or Material trans-4-Methylcyclohexanecarboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 235-959-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only